siRNA Details
Gene Symbol: LOC101927560 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_119374.1 | 3 | 382 | |||
GenBank mRNA | AK097722.1 | - | 3 | 382 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB048258.1 | - | 3 | 382 | Chr. 1 - 83802948 - 83860546 [-] on Build GRCh38 |
