siRNA Details
Gene Symbol: LOC221272 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK025312.1 | - | 1 | 2180 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV657579.1 | - | 2 | 405 | Chr. 6 - 111277876 - 111278744 [+] on Build GRCh38 |
