siRNA Details
Gene Symbol: LOC221272 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK025312.1 | - | 1 | 1287 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG620043.1 | - | 1 | 16 | Chr. 6 - 111277679 - 111278507 [+] on Build GRCh38 |
