siRNA Details
Gene Symbol: LHX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011510105.2 | XP_011508407.1 | 6 | 5834 | ||
XM_011510106.2 | XP_011508408.1 | 6 | 5457 | |||
XM_011510108.2 | XP_011508410.1 | 6 | 5240 | |||
XM_017002755.1 | XP_016858244.1 | 5 | 5952 |
