siRNA Details
Gene Symbol: LHX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011510105.2 | XP_011508407.1 | 6 | 3320 | ||
XM_011510106.2 | XP_011508408.1 | 6 | 2943 | |||
XM_011510108.2 | XP_011508410.1 | 6 | 2726 | |||
XM_017002755.1 | XP_016858244.1 | 5 | 3438 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK026422.1 | - | 1 | 312 | Chr. 1 - 180275950 - 180278837 [+] on Build GRCh38 |
