siRNA Details
Gene Symbol: LINC01539 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040025.1 | 3 | 476 | |||
NR_040026.1 | 2 | 344 | ||||
GenBank mRNA | AK057336.1 | - | 2 | 344 | ||
AK058169.1 | - | 3 | 457 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI829716.1 | - | 227 | Chr. - - [-] on Build |
