siRNA Details
Gene Symbol: LINC01539 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040025.1 | 1 | 186 | |||
NR_040026.1 | 1 | 186 | ||||
GenBank mRNA | AK057336.1 | - | 1 | 186 | ||
AK058169.1 | - | 1 | 167 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI829716.1 | - | 110 | Chr. - - [-] on Build |
