siRNA Details
Gene Symbol: MAPRE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143826.2 | NP_001137298.1 | 8 | 4019 | ||
NM_001143827.2 | NP_001137299.1 | 8 | 4165 | |||
NM_001256420.1 | NP_001243349.1 | 6 | 3979 | |||
NM_014268.3 | NP_055083.1 | 7 | 4107 | |||
NR_046177.1 | 7 | 3922 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CR597075.1 | - | 1 | 1572 | Chr. 18 - 35141741 - 35143462 [+] on Build GRCh38 |
