siRNA Details
Gene Symbol: MAPRE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143826.2 | NP_001137298.1 | 8 | 3339 | ||
NM_001143827.2 | NP_001137299.1 | 8 | 3485 | |||
NM_001256420.1 | NP_001243349.1 | 6 | 3299 | |||
NM_014268.3 | NP_055083.1 | 7 | 3427 | |||
NR_046177.1 | 7 | 3242 |
