siRNA Details
Gene Symbol: MAPRE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143826.2 | NP_001137298.1 | 8 | 4079 | ||
NM_001143827.2 | NP_001137299.1 | 8 | 4225 | |||
NM_001256420.1 | NP_001243349.1 | 6 | 4039 | |||
NM_014268.3 | NP_055083.1 | 7 | 4167 | |||
NR_046177.1 | 7 | 3982 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CR597075.1 | - | 1 | 1632 | Chr. 18 - 35141741 - 35143462 [+] on Build GRCh38 |
