siRNA Details
Gene Symbol: JAML | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK090409.1 | - | 6 | 3454 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK057581.1 | - | 1 | 991 | Chr. 11 - 118199715 - 118201810 [-] on Build GRCh38 |
