siRNA Details
Gene Symbol: HIST1H2BK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK292062.1 | - | 1 | 1106 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX649020.1 | - | 1 | 132 | Chr. 6 - 27142092 - 27145881 [-] on Build GRCh38 |
