siRNA Details
Gene Symbol: LOC344887 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033752.2 | 6 | 1642 | |||
GenBank mRNA | BX640843.1 | - | 1 | 3499 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC344887 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033752.2 | 6 | 1642 | |||
GenBank mRNA | BX640843.1 | - | 1 | 3499 |