siRNA Details
Gene Symbol: RNF220 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK124790.1 | - | 1 | 208 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK124261.1 | - | 1 | 1710 | Chr. 1 - 44641631 - 44643422 [+] on Build GRCh38 |
