siRNA Details
Gene Symbol: STAT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011511705.2 | XP_011510007.1 | 4 | 334 | ||
GenBank mRNA | EU304791.1 | - | 4 | 321 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK001864.1 | - | 1 | 727 | Chr. 2 - 191154118 - 191156070 [-] on Build GRCh38 |
