siRNA Details
Gene Symbol: SDCBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK022318.1 | - | 1 | 152 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK091587.1 | - | 1 | 101 | Chr. 2 - 201174377 - 201176688 [+] on Build GRCh38 |
