siRNA Details
Gene Symbol: LOC101928505 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC020897.1 | - | 1 | 706 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG567427.1 | - | 1 | 709 | Chr. 5 - 57669838 - 57670640 [+] on Build GRCh38 |
