siRNA Details
Gene Symbol: LOC101928505 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104668.1 | 3 | 765 | |||
GenBank mRNA | BC020897.1 | - | 1 | 254 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG567427.1 | - | 1 | 255 | Chr. 5 - 57669838 - 57670640 [+] on Build GRCh38 |
