siRNA Details
Gene Symbol: LOC101928505 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104668.1 | 3 | 1310 | |||
GenBank mRNA | BC020897.1 | - | 1 | 799 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101928505 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104668.1 | 3 | 1310 | |||
GenBank mRNA | BC020897.1 | - | 1 | 799 |