siRNA Details
Gene Symbol: LOC100506446 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK092589.1 | - | 2, 3 | 350 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA869734.1 | - | 2, 3 | 350 | Chr. 14 - 49943870 - 49961960 [-] on Build GRCh38 |
