siRNA Details
Gene Symbol: LINC00587 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103830.1 | 5 | 508 | |||
GenBank mRNA | BC038565.2 | - | 5 | 545 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00587 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103830.1 | 5 | 508 | |||
GenBank mRNA | BC038565.2 | - | 5 | 545 |