siRNA Details
Gene Symbol: LOC101928708 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110939.1 | 3 | 522 | |||
GenBank mRNA | BC043543.1 | - | 3 | 515 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI829296.1 | - | 3 | 515 | Chr. 16 - 87212122 - 87225816 [+] on Build GRCh38 |
