siRNA Details
Gene Symbol: GS1-24F4.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045217.1 | 3 | 519 | |||
GenBank mRNA | BC043547.1 | - | 3 | 519 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CD557887.1 | - | 1 | 184 | Chr. 8 - 6841693 - 6842571 [+] on Build GRCh38 |
