siRNA Details
Gene Symbol: 4930432F03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015285.1 | - | 1 | 536 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714842.1 | - | 1 | 535 | Chr. 4 - 59002338 - 59003075 [-] on Build GRCm38 |
