siRNA Details
Gene Symbol: Gm17344 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK038784.1 | - | 2 | 267 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB632057.1 | - | 2 | 267 | Chr. 8 - 109862187 - 109868455 [-] on Build GRCm38 |
