siRNA Details
Gene Symbol: Gm15179 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037976.1 | 1, 2 | 133 | |||
GenBank mRNA | AK039780.1 | - | 1, 2 | 131 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB634114.1 | - | 1, 2 | 131 | Chr. 1 - 75368212 - 75375012 [-] on Build GRCm38 |
