siRNA Details
Gene Symbol: Gm16892 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK149401.1 | - | 3 | 212 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CJ133848.1 | - | 3 | 212 | Chr. 16 - 57167401 - 57173375 [+] on Build GRCm38 |
