siRNA Details
Gene Symbol: Gm5091 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046164.1 | 3 | 223 | |||
GenBank mRNA | AK082876.1 | - | 3 | 224 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY728917.1 | - | 3 | 224 | Chr. 17 - 15232135 - 15240783 [-] on Build GRCm38 |
