siRNA Details
Gene Symbol: 1700055D16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006793.1 | - | 1 | 58 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706775.1 | - | 1 | 54 | Chr. 6 - 134792363 - 134820678 [+] on Build GRCm38 |
