siRNA Details
Gene Symbol: 4933432I03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045657.1 | 2 | 114 | |||
GenBank mRNA | AK017023.1 | - | 2 | 115 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY716707.1 | - | 2 | 115 | Chr. 14 - 102987570 - 103033803 [-] on Build GRCm38 |
