siRNA Details
Gene Symbol: 2900009J20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK013507.1 | - | 1 | 1779 | ||
AK045450.1 | - | 1 | 2120 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB672254.1 | - | 1 | 187 | Chr. 12 - 72799414 - 72799818 [+] on Build GRCm38 |
