siRNA Details
Gene Symbol: 2900009J20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK013507.1 | - | 1 | 155 | ||
AK045450.1 | - | 1 | 496 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 2900009J20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK013507.1 | - | 1 | 155 | ||
AK045450.1 | - | 1 | 496 |