siRNA Details
Gene Symbol: 4933407O12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016733.1 | - | 1 | 496 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY716414.1 | - | 1 | 496 | Chr. 17 - 80062801 - 80063735 [+] on Build GRCm38 |
