siRNA Details
Gene Symbol: 4933408J17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045810.1 | 2 | 425 | |||
GenBank mRNA | AK016739.1 | - | 2 | 428 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY716420.1 | - | 2 | 428 | Chr. 10 - 93589419 - 93596255 [+] on Build GRCm38 |
