siRNA Details
Gene Symbol: 1700109I08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045936.1 | 1 | 17 | |||
GenBank mRNA | AK007151.1 | - | 1 | 19 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY707235.1 | - | 1 | 18 | Chr. 14 - 39655663 - 39685293 [+] on Build GRCm38 |
