siRNA Details
Gene Symbol: 1700110C19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045461.1 | 3 | 359 | |||
GenBank mRNA | AK007157.1 | - | 3 | 360 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CA464730.1 | - | 3 | 325 | Chr. 17 - 10324602 - 10329340 [+] on Build GRCm38 |
