siRNA Details
Gene Symbol: 1700109K24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108037.1 | 3 | 681 | |||
GenBank mRNA | AK007154.1 | - | 3 | 636 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY707238.1 | - | 3 | 635 | Chr. 15 - 77084443 - 77096279 [+] on Build GRCm38 |
