siRNA Details
Gene Symbol: Osbpl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK004353.1 | - | 2 | 456 | ||
AK157944.1 | - | 1 | 1236 | |||
BY704583.1 | - | 2 | 456 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Osbpl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK004353.1 | - | 2 | 456 | ||
AK157944.1 | - | 1 | 1236 | |||
BY704583.1 | - | 2 | 456 |