siRNA Details
Gene Symbol: 2010320H13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK008591.1 | - | 3 | 311 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY708483.1 | - | 3 | 307 | Chr. 5 - 44099648 - 44101954 [-] on Build GRCm38 |
