siRNA Details
Gene Symbol: 9630009A06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020673.1 | - | 396 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY722409.1 | - | 2 | 396 | Chr. 15 - 30961455 - 30962878 [-] on Build GRCm38 |
