siRNA Details
Gene Symbol: B230110G15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020962.1 | - | 4 | 634 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY727005.1 | - | 4 | 634 | Chr. 6 - 138580624 - 138599213 [+] on Build GRCm38 |
