siRNA Details
Gene Symbol: B230112I24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020969.1 | - | 1 | 563 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY727025.1 | - | 1 | 563 | Chr. 6 - 45073566 - 45074215 [+] on Build GRCm38 |
