siRNA Details
Gene Symbol: Gm15509 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK133047.1 | - | 3 | 2061 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB016497.2 | - | 3 | 523 | Chr. 1 - 187120869 - 187122820 [-] on Build GRCm38 |
