siRNA Details
Gene Symbol: Gm12440 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110516.1 | 2 | 717 | |||
GenBank mRNA | AK076933.1 | - | 2 | 718 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB016323.2 | - | 1 | 262 | Chr. 3 - 99052662 - 99053730 [+] on Build GRCm38 |
