siRNA Details
Gene Symbol: Gm12440 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110516.1 | 2 | 436 | |||
GenBank mRNA | AK076933.1 | - | 2 | 437 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB615799.1 | - | 2 | 437 | Chr. 3 - 99049822 - 99052802 [+] on Build GRCm38 |
