siRNA Details
Gene Symbol: 4933407I05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040731.1 | 3 | 509 | |||
GenBank mRNA | AK076937.1 | - | 3 | 510 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715817.1 | - | 3 | 510 | Chr. 9 - 51914429 - 51926841 [+] on Build GRCm38 |
