siRNA Details
Gene Symbol: Enpp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014707.1 | - | 1 | 790 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CJ257496.1 | - | 1 | 440 | Chr. 10 - 24637920 - 24638466 [-] on Build GRCm38 |
