siRNA Details
Gene Symbol: Gm13546 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045895.1 | 3 | 247 | |||
GenBank mRNA | AK041746.1 | - | 3 | 221 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB638452.1 | - | 3 | 218 | Chr. 2 - 58164003 - 58176810 [+] on Build GRCm38 |
