siRNA Details
Gene Symbol: Gm13648 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC048384.1 | - | 2 | 455 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BQ715864.1 | - | 2 | 454 | Chr. 4 - 156053373 - 156059801 [-] on Build GRCm38 |
