siRNA Details
Gene Symbol: Gm41669 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK082205.1 | - | 3 | 536 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB650013.1 | - | 3 | 533 | Chr. 18 - 13286584 - 13306336 [-] on Build GRCm38 |
